Assay Details
Target Gene Details
Entrez Gene ID: | 58198 |
Gene Name: | sal-like 1 (Drosophila) |
Gene Aliases: |
Msal-3 |
Location: |
Chr.8:89027239-89044162 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sall1 | NM_021390.3 | NP_067365.2 | ||
| BC062937.1 | AAH62937.1 |
More Information
Set Membership: |
|

Genomic Map