Assay Details
Target Gene Details
Entrez Gene ID: | 244550 |
Gene Name: | podocan-like 1 |
Gene Aliases: |
5832418A03Rik, Gm506 |
Location: |
Chr.8:84125927-84143016 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Podnl1 | NM_001013384.2 | 1 | 56 | NP_001013402.2 |
| BC063764.1 | 1 | 56 | AAH63764.1 | |
| DQ303463.1 |

Genomic Map