Assay Details
Target Gene Details
Entrez Gene ID: | 18970 |
Gene Name: | polymerase (DNA directed), beta |
Gene Aliases: |
A430088C08Rik |
Location: |
Chr.8:22628119-22653468 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polb | NM_011130.2 | NP_035260.1 | ||
| AF513911.1 | AAM49616.1 | |||
| AK077127.1 | BAC36630.1 | |||
| AK146745.1 | BAE27405.1 | |||
| AK151436.1 | BAE30399.1 | |||
| AV488131.1 | ||||
| BC006681.1 | AAH06681.1 | |||
| BC060998.1 | AAH60998.1 |
More Information
Set Membership: |
|

Genomic Map