Assay Details
Target Gene Details
Entrez Gene ID: | 67789 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
6330580J24Rik, C77829, mir-425 |
Location: |
Chr.9:108567496-108572773 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dalrd3 | NM_026378.3 | 1 | 130 | NP_080654.3 |
| AK002439.1 | 1 | 131 | BAC24990.1 | |
| AK149963.1 | 1 | 131 | BAE29197.1 | |
| AK169405.1 | 1 | 128 | BAE41151.1 | |
| AK171818.1 | 1 | 131 | BAE42679.1 | |
| BC013483.1 | 1 | 111 | AAH13483.2 |

Genomic Map