Assay Details
Target Gene Details
Entrez Gene ID: | 66113 |
Gene Name: | apolipoprotein A-V |
Gene Aliases: |
1300007O05Rik, Apoav, RAP3 |
Location: |
Chr.9:46268608-46271919 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoa5 |

Genomic Map