Assay Details
Target Gene Details
Entrez Gene ID: | 270152 |
Gene Name: | adhesion molecule, interacts with CXADR antigen 1 |
Gene Aliases: |
AMICA, Crea7, Gm638 |
Location: |
Chr.9:45079162-45108531 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amica1 | NM_001005421.4 | NP_001005421.3 | ||
| AA120429.1 | ||||
| AY093688.1 | AAM15732.1 | |||
| BC050133.1 | AAH50133.1 |
More Information
Set Membership: |
|

Genomic Map