Assay Details
Target Gene Details
Entrez Gene ID: | 102632 |
Gene Name: | acyl-Coenzyme A dehydrogenase family, member 11 |
Gene Aliases: |
5730439E10Rik, AI987948 |
Location: |
Chr.9:104063703-104127646 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acad11 | NM_175324.3 | NP_780533.2 | ||
| AK077531.1 | BAC36849.2 | |||
| AK128998.1 | ||||
| AK139918.1 | ||||
| AK145561.1 | BAE26511.1 | |||
| AK145834.1 | BAE26684.1 | |||
| BC045199.1 | AAH45199.1 |
Target Gene Details
Entrez Gene ID: | 252837 |
Gene Name: | atypical chemokine receptor 4 |
Gene Aliases: |
A630091E18Rik, CCBP2, CCR11, CCX-CKR, CCX-CKR1, Ccrl1, Cmkbrl1, PPR1, VSHK1 |
Location: |
Chr.9:104097682-104126643 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ackr4 | NM_145700.2 | NP_663746.2 | ||
| AY072938.1 | 1 | 318 | AAL68962.1 | |
| BC140999.1 |
More Information
Set Membership: |
|

Genomic Map