Assay Details
Target Gene Details
Entrez Gene ID: | 54725 |
Gene Name: | cell adhesion molecule 1 |
Gene Aliases: |
2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1 |
Location: |
Chr.9:47530239-47853385 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadm1 | NM_001025600.1 | 7 | 895 | NP_001020771.1 |
| NM_018770.3 | 7 | 895 | NP_061240.3 | |
| NM_207675.2 | 7 | 895 | NP_997558.2 | |
| NM_207676.2 | 7 | 895 | NP_997559.1 | |
| AB021964.1 | 4 | 424 | BAA87914.1 | |
| AB021965.1 | 4 | 424 | BAA87915.1 | |
| AB021966.1 | 4 | 424 | BAA87916.1 | |
| AB021967.1 | 4 | 424 | BAA87917.1 | |
| AB052293.1 | 7 | 895 | BAB60686.1 | |
| AB064265.2 | 7 | 874 | BAB83501.2 | |
| AB092414.1 | 7 | 874 | BAC66173.1 | |
| AB183399.1 | 7 | 874 | BAD30018.1 | |
| AB183400.1 | 7 | 874 | BAD30019.1 | |
| AB183401.1 | 7 | 874 | BAD30020.1 | |
| AB183402.1 | 7 | 874 | BAD30021.1 | |
| AB377192.1 | 7 | 895 | ||
| AF061260.1 | 6 | 732 | AAC67243.1 | |
| AF434663.1 | 7 | 990 | AAL86736.1 | |
| AF539424.1 | 7 | 887 | AAN01614.1 | |
| AK013775.1 | 7 | 896 | ||
| AK013911.1 | 7 | 989 | ||
| AK053839.1 | 7 | 896 | ||
| AY351388.1 | 7 | 874 | AAQ02381.1 | |
| BC057125.1 | 7 | 883 | AAH57125.1 | |
| BC090970.1 | 7 | 1074 | AAH90970.1 | |
| BC094661.1 | 4 | 445 | AAH94661.1 | |
| BC095986.1 | 7 | 887 | AAH95986.1 | |
| DQ279856.1 | 7 | 874 | ||
| EU541474.1 | 1 | 44 | ||
| EU541475.1 | 1 | 44 | ||
| EU541476.1 | 1 | 44 | ||
| EU541477.1 | 1 | 44 | ||
| EU541478.1 | 1 | 44 | ||
| JQ782219.1 | 7 | 874 |
Target Gene Details
Entrez Gene ID: | 100038460 |
Gene Name: | predicted gene 10677 |
Gene Aliases: |
ENSMUSG00000074363 |
Location: |
Chr.9:47799160-47818208 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10677 | NR_046048.1 | |||
| AK048734.1 | BAE20665.1 |
More Information
Set Membership: |
|

Genomic Map