Assay Details
Target Gene Details
Entrez Gene ID: | 100529079 |
Gene Name: | RIKEN cDNA 5830418P13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:103423578-103461260 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5830418P13Rik | NR_040466.1 | |||
| AK134047.1 |
Target Gene Details
Entrez Gene ID: | 107993 |
Gene Name: | beaded filament structural protein 2, phakinin |
Gene Aliases: |
AI448593, CP49 |
Location: |
Chr.9:103424924-103480414 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bfsp2 | NM_001002896.2 | 7 | 1461 | NP_001002896.1 |
| AK134810.1 | 7 | 1393 | BAE22294.1 | |
| BC062815.1 | 5 | 663 | AAH62815.1 | |
| BC068172.1 | 7 | 1461 | AAH68172.1 | |
| S55549.1 | 5 | 770 | AAB25419.1 |
More Information
Set Membership: |
|

Genomic Map