Assay Details
Target Gene Details
Entrez Gene ID: | 13627 |
Gene Name: | eukaryotic translation elongation factor 1 alpha 1 |
Gene Aliases: |
- |
Location: |
Chr.9:78478453-78481724 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
More Information
Set Membership: |
|

Genomic Map