Assay Details
Target Gene Details
Entrez Gene ID: | 74602 |
Gene Name: | RIKEN cDNA 4833428L15 gene |
Gene Aliases: |
- |
Location: |
Chr.9:45416626-45431232 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4833428L15Rik | NR_040732.1 | |||
| AK014786.1 |
Target Gene Details
Entrez Gene ID: | 114873 |
Gene Name: | Down syndrome cell adhesion molecule like 1 |
Gene Aliases: |
4921507G06Rik, 4930435C18Rik, BB239540, mKIAA1132 |
Location: |
Chr.9:45429495-45753713 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dscaml1 | NM_001081270.1 | NP_001074739.1 | ||
| AF487345.1 | AAL99983.1 | |||
| AF487347.1 | AAL99985.1 | |||
| AK147531.1 |
More Information
Set Membership: |
|

Genomic Map