Assay Details
Target Gene Details
Entrez Gene ID: | 13663 |
Gene Name: | etoposide induced 2.4 mRNA |
Gene Aliases: |
AA536736, AI115355, PIG8 |
Location: |
Chr.9:36779153-36797393 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ei24 | NM_001199494.1 | 3 | 364 | NP_001186423.1 |
| NM_007915.5 | 3 | 248 | NP_031941.1 | |
| AK145018.1 | 3 | 325 | ||
| AK147776.1 | 3 | 277 | BAE28130.1 | |
| AK159927.1 | 3 | 306 | BAE35489.1 | |
| AK172233.1 | 3 | 328 | BAE42899.1 | |
| AK184366.1 | ||||
| AK188309.1 | ||||
| AK206494.1 | ||||
| BU148129.1 | 3 | 317 | ||
| BY282890.1 | 3 | 365 | ||
| U41751.2 | 3 | 252 | AAC52483.2 |

Genomic Map