Assay Details
Target Gene Details
Entrez Gene ID: | 64655 |
Gene Name: | mitochondrial ribosomal protein S22 |
Gene Aliases: |
3100002P07Rik, Rpms22 |
Location: |
Chr.9:98588730-98601679 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps22 | NM_025485.3 | 3 | 421 | NP_079761.1 |
| AK003836.1 | 2 | 189 | BAB23027.2 | |
| AK013925.1 | 3 | 402 | BAB29061.1 | |
| AK154600.1 | 3 | 402 | BAE32703.1 | |
| BC051198.1 | 3 | 401 | AAH51198.1 | |
| CJ183743.1 |
More Information
Set Membership: |
|

Genomic Map