Assay Details
Target Gene Details
Entrez Gene ID: | 54369 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 |
Gene Aliases: |
nm23-M6 |
Location: |
Chr.9:109832711-109842974 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme6 | AK004201.1 |

Genomic Map