Assay Details
Target Gene Details
Entrez Gene ID: | 12091 |
Gene Name: | galactosidase, beta 1 |
Gene Aliases: |
AW125515, Bge, Bgl, Bgl-e, Bgl-s, Bgl-t, Bgs, Bgt, C130097A14Rik |
Location: |
Chr.9:114401078-114474379 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glb1 | NM_009752.2 | NP_033882.1 | ||
| NR_108101.1 | ||||
| NR_108102.1 | ||||
| AK034049.1 | ||||
| AK042069.1 | ||||
| AK048460.1 | BAC33344.1 | |||
| AK076517.1 | BAC36375.1 | |||
| AK153218.1 | ||||
| AK166473.1 | ||||
| AK167271.1 | ||||
| AK171593.1 | BAE42549.1 | |||
| AK171760.1 | ||||
| BC028875.1 | AAH28875.1 | |||
| M57734.1 | AAA37293.1 |
Target Gene Details
Entrez Gene ID: | 100504715 |
Gene Name: | transmembrane protein with metallophosphoesterase domain |
Gene Aliases: |
- |
Location: |
Chr.9:114401095-114411201 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmppe | NM_001200002.1 | NP_001186931.1 | ||
| AK046107.1 | ||||
| CN705942.1 |
More Information
Set Membership: |
|

Genomic Map