Assay Details
Target Gene Details
Entrez Gene ID: | 64655 |
Gene Name: | mitochondrial ribosomal protein S22 |
Gene Aliases: |
3100002P07Rik, Rpms22 |
Location: |
Chr.9:98588730-98601679 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps22 | NM_025485.3 | NP_079761.1 | ||
| AK003836.1 | BAB23027.2 | |||
| AK013925.1 | BAB29061.1 | |||
| AK154600.1 | BAE32703.1 | |||
| BC021882.1 | AAH21882.1 | |||
| BC051198.1 | AAH51198.1 | |||
| BE983156.1 |
More Information
Set Membership: |
|

Genomic Map