Assay Details
Target Gene Details
Entrez Gene ID: | 69524 |
Gene Name: | endothelial cell-specific adhesion molecule |
Gene Aliases: |
2310008D05Rik, Esam1, W117m |
Location: |
Chr.9:37528055-37538319 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esam | NM_027102.3 | 4 | 670 | NP_081378.1 |
| AF361882.1 | 4 | 655 | AAK51504.1 | |
| AK009223.1 | 4 | 646 | ||
| AK156399.1 | 4 | 668 | BAE33699.1 | |
| BC019460.1 | 4 | 654 | ||
| BC049249.1 | 4 | 650 | AAH49249.1 | |
| BG077536.2 | 2 | 104 | ||
| BQ043033.1 | 4 | 611 |

Genomic Map