Assay Details
Target Gene Details
Entrez Gene ID: | 17386 |
Gene Name: | matrix metallopeptidase 13 |
Gene Aliases: |
Clg, MMP-13, Mmp1 |
Location: |
Chr.9:7272514-7283333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp13 | NM_008607.2 | NP_032633.1 | ||
| AK150728.1 | BAE29805.1 | |||
| AK151626.1 | BAE30561.1 | |||
| AK152507.1 | BAE31273.1 | |||
| AK152916.1 | BAE31593.1 | |||
| AK153458.1 | BAE32010.1 | |||
| AK155188.1 | BAE33104.1 | |||
| AK159921.1 | BAE35483.1 | |||
| AK160819.1 | BAE36033.1 | |||
| BC125320.1 | ||||
| BC125322.1 | ||||
| X66473.1 | CAA47102.1 |
More Information
Set Membership: |
|

Genomic Map