Assay Details
Target Gene Details
Entrez Gene ID: | 74840 |
Gene Name: | mesencephalic astrocyte-derived neurotrophic factor |
Gene Aliases: |
3230402M22Rik, AA407711, AA408789, AA673178, Armet, D17914, D18Mgi17 |
Location: |
Chr.9:106868310-106891985 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Manf | NM_029103.3 | NP_083379.2 | ||
| AB009291.1 | ||||
| AK014338.1 | BAB29281.1 | |||
| AK131997.1 | BAE20927.1 | |||
| AK165646.1 | BAE38314.1 | |||
| AK167580.1 | ||||
| AK176996.1 | ||||
| AK186239.1 |

Genomic Map