Assay Details
Target Gene Details
Entrez Gene ID: | 83995 |
Gene Name: | matrix metallopeptidase 1a (interstitial collagenase) |
Gene Aliases: |
Mcol-A, Mcola |
Location: |
Chr.9:7464141-7476869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp1a | NM_032006.3 | NP_114395.1 | ||
| AJ278462.1 | CAC18880.1 | |||
| AK049552.1 | BAC33807.1 | |||
| BC117756.2 |
More Information
Set Membership: |
|

Genomic Map