Assay Details
Target Gene Details
Entrez Gene ID: | 83996 |
Gene Name: | matrix metallopeptidase 1b (interstitial collagenase) |
Gene Aliases: |
Mcol-B, Mcolb |
Location: |
Chr.9:7367640-7388026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp1b | NM_032007.3 | NP_114396.3 | ||
| AJ278461.1 | CAC18879.1 |
More Information
Set Membership: |
|

Genomic Map