Assay Details
Target Gene Details
Entrez Gene ID: | 546143 |
Gene Name: | cell cycle progression 1, opposite strand |
Gene Aliases: |
EG546143, Gm5918 |
Location: |
Chr.9:72979717-72985376 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccpg1os | NM_001198789.1 | 1 | 131 | NP_001185718.1 |
| AK145722.1 | 1 | 137 |

Genomic Map