Assay Details
Target Gene Details
Entrez Gene ID: | 74770 |
Gene Name: | hedgehog acyltransferase-like |
Gene Aliases: |
1110011D13Rik, Gup1 |
Location: |
Chr.9:121784016-121792525 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hhatl | NM_029095.2 | 12 | 1715 | NP_083371.2 |
| NR_027967.1 | 12 | 1595 | ||
| AK003605.1 | 12 | 1592 | BAB22885.1 | |
| AK157924.1 | 12 | 1513 | BAE34266.1 | |
| BC024464.1 | 12 | 1571 | AAH24464.1 |
Target Gene Details
Entrez Gene ID: | 72330 |
Gene Name: | kelch-like 40 |
Gene Aliases: |
2310024D23Rik, Kbtbd5 |
Location: |
Chr.9:121777101-121791002 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl40 |
More Information
Set Membership: |
|

Genomic Map