Assay Details
Target Gene Details
Entrez Gene ID: | 102423 |
Gene Name: | histone H4 transcription factor |
Gene Aliases: |
AA589481, HiNF-P, Mizf |
Location: |
Chr.9:44292438-44305705 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hinfp | NM_172162.3 | NP_751894.2 | ||
| AK049348.1 | ||||
| AK049633.1 | BAC33849.1 | |||
| BC029091.1 | AAH29091.1 | |||
| CT010239.1 | CAJ18447.1 |
More Information
Set Membership: |
|

Genomic Map