Assay Details
Target Gene Details
Entrez Gene ID: | 17392 |
Gene Name: | matrix metallopeptidase 3 |
Gene Aliases: |
SLN-1, SLN1, STR-1, Stmy1, Str1 |
Location: |
Chr.9:7445822-7455975 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp3 | NM_010809.1 | NP_034939.1 | ||
| AK148467.1 | BAE28570.1 | |||
| BC006725.1 | AAH06725.1 | |||
| X63162.1 | CAA44860.1 | |||
| X66402.1 | CAA47029.1 |
More Information
Set Membership: |
|

Genomic Map