Assay Details
Target Gene Details
Entrez Gene ID: | 235169 |
Gene Name: | FAD-dependent oxidoreductase domain containing 1 |
Gene Aliases: |
BC024806, TEG-23, Tex23 |
Location: |
Chr.9:35204208-35211055 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxred1 | NM_001291448.1 | NP_001278377.1 | ||
| NM_001291449.1 | NP_001278378.1 | |||
| NM_172291.2 | NP_758495.1 | |||
| AK054231.1 | BAC35698.1 | |||
| AK144759.1 | BAE26052.1 | |||
| AK163722.1 | BAE37472.1 | |||
| BC024806.1 | AAH24806.1 |
More Information
Set Membership: |
|

Genomic Map