Assay Details
Target Gene Details
Entrez Gene ID: | 665268 |
Gene Name: | ribosomal protein L17 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.9:58202897-58208808 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1600029O15Rik | NR_033522.1 | |||
| AK005564.2 | BAB24124.2 |
Target Gene Details
Entrez Gene ID: | 320563 |
Gene Name: | immunoglobulin superfamily containing leucine-rich repeat 2 |
Gene Aliases: |
B930052A04Rik, Linx, Mbu-3, mKIAA1465 |
Location: |
Chr.9:58196297-58204319 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Islr2 | NM_001161541.1 | NP_001155013.1 | ||
| BB611833.1 |
More Information
Set Membership: |
|

Genomic Map