Assay Details
Target Gene Details
Entrez Gene ID: | 17427 |
Gene Name: | meiosis-specific nuclear structural protein 1 |
Gene Aliases: |
AW546487 |
Location: |
Chr.9:72438518-72458576 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mns1 | NM_008613.3 | 2 | 306 | NP_032639.1 |
| AK013048.1 | 2 | 273 | ||
| AK132772.1 | 2 | 281 | ||
| BC046624.1 | 2 | 219 | AAH46624.1 | |
| D14849.1 | 2 | 337 | BAA03577.1 |
More Information
Set Membership: |
|

Genomic Map