Assay Details
Target Gene Details
Entrez Gene ID: | 15561 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 3A |
Gene Aliases: |
5-HT3 |
Location: |
Chr.9:48899213-48911151 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htr3a | NM_001099644.1 | NP_001093114.1 | ||
| NM_013561.2 | NP_038589.2 | |||
| AF513221.1 | AAM47014.1 | |||
| AY605711.1 | AAT37716.1 | |||
| BC144799.1 | ||||
| BC145743.1 | ||||
| M74425.1 | AAA37124.1 | |||
| X72395.1 | CAA51089.1 |
More Information
Set Membership: |
|

Genomic Map