Assay Details
Target Gene Details
Entrez Gene ID: | 15561 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 3A |
Gene Aliases: |
5-HT3 |
Location: |
Chr.9:48899213-48911151 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htr3a | NM_001099644.1 | 8 | 1134 | NP_001093114.1 |
| NM_013561.2 | 8 | 1134 | NP_038589.2 | |
| AF513221.1 | 8 | 1011 | AAM47014.1 | |
| AY605711.1 | 8 | 1000 | AAT37716.1 | |
| BC144799.1 | 8 | 1056 | ||
| BC145743.1 | 8 | 1056 | ||
| M74425.1 | 8 | 1054 | AAA37124.1 | |
| X72395.1 | 8 | 1018 | CAA51089.1 |
More Information
Set Membership: |
|

Genomic Map