Assay Details
Target Gene Details
Entrez Gene ID: | 17083 |
Gene Name: | transmembrane emp24 domain containing 1 |
Gene Aliases: |
Il1rl1l, Ly84l, St2l |
Location: |
Chr.9:21507380-21510187 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmed1 | NM_010744.3 | NP_034874.2 | ||
| AK133513.1 | BAE21696.1 | |||
| AK160525.1 | BAE35843.1 | |||
| AK166092.1 | ||||
| BC006715.1 | ||||
| BC138498.1 | ||||
| BC138499.1 | ||||
| U41805.1 | AAC52472.1 |
More Information
Set Membership: |
|

Genomic Map