Assay Details
Target Gene Details
Entrez Gene ID: | 208084 |
Gene Name: | PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae) |
Gene Aliases: |
4631410M14, AI449441 |
Location: |
Chr.9:65587147-65595967 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pif1 | NM_172453.3 | NP_766041.1 | ||
| AK028460.1 | BAC25963.1 | |||
| AK049353.1 | BAC33702.1 | |||
| AK154072.1 | ||||
| AY100322.1 | AAM50051.1 | |||
| AY498715.1 | AAS77397.1 | |||
| BC046611.1 | AAH46611.1 |
More Information
Set Membership: |
|

Genomic Map