Assay Details
Target Gene Details
Entrez Gene ID: | 71687 |
Gene Name: | transmembrane protein 25 |
Gene Aliases: |
0610039J01Rik, AI429491 |
Location: |
Chr.9:44793772-44799273 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem25 | NM_027865.2 | NP_082141.1 | ||
| AK002841.1 | BAB22398.1 | |||
| AK090375.1 | BAC41190.1 | |||
| AK154206.1 | BAE32437.1 | |||
| AK165199.1 | BAE38074.1 | |||
| AK165827.1 | BAE38398.1 | |||
| BC060057.1 | AAH60057.1 | |||
| BC060243.1 | AAH60243.1 |

Genomic Map