Assay Details
Target Gene Details
Entrez Gene ID: | 19035 |
Gene Name: | peptidylprolyl isomerase B |
Gene Aliases: |
AA408962, AA553318, AI844835, Cphn-2, Cphn2, CyP-20b |
Location: |
Chr.9:66060169-66066629 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppib | NM_011149.2 | NP_035279.2 | ||
| AK002357.1 | BAB22036.1 | |||
| AK128977.1 | BAC87663.1 | |||
| AK154073.1 | BAE32357.1 | |||
| AL363449.1 | ||||
| BC013061.1 | AAH13061.1 | |||
| M60456.1 | AAA37498.1 | |||
| X58990.1 | CAA41736.1 |
More Information
Set Membership: |
|

Genomic Map