Assay Details
Target Gene Details
Entrez Gene ID: | 66712 |
Gene Name: | sperm equatorial segment protein 1 |
Gene Aliases: |
4921508E09Rik, AI428149 |
Location: |
Chr.9:62270729-62282179 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spesp1 | NM_025721.2 | 1 | 136 | NP_079997.1 |
| AK014843.1 | 1 | 137 | BAB29577.1 | |
| AK015620.1 | 1 | 167 | BAB29908.1 | |
| BC050754.1 | 1 | 144 | AAH50754.1 |

Genomic Map