Assay Details
Target Gene Details
Entrez Gene ID: | 19944 |
Gene Name: | ribosomal protein L29 |
Gene Aliases: |
Rpl43 |
Location: |
Chr.9:106429432-106431568 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl29 | NM_009082.2 | NP_033108.1 | ||
| AK013307.1 | BAB28782.1 | |||
| AK088551.1 | BAC40419.1 | |||
| AK141477.1 | BAE24697.1 | |||
| AK167284.1 | BAE39390.1 | |||
| BC081467.1 | AAH81467.1 | |||
| BC082292.1 | AAH82292.1 | |||
| BC086897.1 | AAH86897.1 | |||
| BC086898.1 | AAH86898.1 | |||
| BC087950.1 | AAH87950.1 | |||
| BC092262.1 | AAH92262.1 | |||
| BC094039.1 | AAH94039.1 | |||
| BC096575.1 | AAH96575.1 | |||
| L08651.1 | AAA16857.1 |
More Information
Set Membership: |
|

Genomic Map