Assay Details
Target Gene Details
Entrez Gene ID: | 56032 |
Gene Name: | nitrogen permease regulator-like 2 |
Gene Aliases: |
2810446G01Rik, G21, NPR2L, Tusc4 |
Location: |
Chr.9:107542206-107545706 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nprl2 | NM_018879.2 | 11 | 1306 | NP_061367.1 |
| AF131207.1 | 11 | 1305 | AAD33474.1 | |
| AI843290.1 | 1 | 168 | ||
| AK010331.1 | 11 | 1224 | BAB26858.1 | |
| AK013299.1 | 11 | 1268 | BAB28777.1 | |
| AK013691.1 | 11 | 1257 | ||
| AK088925.1 | 9 | 1596 | ||
| BC026548.1 | 11 | 1222 | AAH26548.1 |
More Information
Set Membership: |
|

Genomic Map