Assay Details
Target Gene Details
Entrez Gene ID: | 56032 |
Gene Name: | nitrogen permease regulator-like 2 |
Gene Aliases: |
2810446G01Rik, G21, NPR2L, Tusc4 |
Location: |
Chr.9:107542206-107545706 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nprl2 | NM_018879.2 | NP_061367.1 | ||
| AF131207.1 | AAD33474.1 | |||
| AK010331.1 | BAB26858.1 | |||
| AK013299.1 | BAB28777.1 | |||
| AK013691.1 | ||||
| AK088925.1 | 7 | 866 | ||
| BC026548.1 | AAH26548.1 |
More Information
Set Membership: |
|

Genomic Map