Assay Details
Target Gene Details
Entrez Gene ID: | 272643 |
Gene Name: | protease, serine 43 |
Gene Aliases: |
Gm661, Tessp3 |
Location: |
Chr.9:110826690-110831507 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss43 | NM_199471.1 | 5 | 1141 | NP_955765.1 |
| AB100999.1 | 5 | 1141 | BAD06396.1 |
More Information
Set Membership: |
|

Genomic Map