Assay Details
Target Gene Details
Entrez Gene ID: | 22688 |
Gene Name: | zinc finger protein 26 |
Gene Aliases: |
5033428C05Rik, KRAB15, Zfp-26, Zfp70, Zfp81-rs1, mKIAA4196, mkr-3, mszf14, mszf52 |
Location: |
Chr.9:20428318-20460169 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp26 | NM_011753.3 | NP_035883.2 | ||
| AK085739.1 | ||||
| CA871137.1 |
More Information
Set Membership: |
|

Genomic Map