Assay Details
Target Gene Details
Entrez Gene ID: | 235028 |
Gene Name: | zinc finger protein 426 |
Gene Aliases: |
2900057C04Rik, KRAB1, Zfo61, Zfp68-rs1, Znf426, mKIAA4237 |
Location: |
Chr.9:20468548-20493922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp426 | NM_001110309.1 | NP_001103779.1 | ||
| NM_146221.4 | NP_666333.1 | |||
| AK036912.1 | BAC29636.1 | |||
| AK085144.1 | BAC39374.1 | |||
| AK220563.1 | BAD90329.1 | |||
| BC023931.1 | AAH23931.1 | |||
| BC024805.1 | AAH24805.1 |

Genomic Map