Assay Details
Target Gene Details
Entrez Gene ID: | 321006 |
Gene Name: | Vpr (HIV-1) binding protein |
Gene Aliases: |
AI447437, B930007L02Rik, mKIAA0800 |
Location: |
Chr.9:106814172-106880992 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vprbp | NM_001015507.2 | 18 | 3847 | NP_001015507.1 |
| AK122372.1 | 18 | 3810 | BAC65654.3 | |
| AK135721.1 | BAE22628.1 | |||
| BC065119.1 | 18 | 3847 | AAH65119.1 |

Genomic Map