Assay Details
Target Gene Details
Entrez Gene ID: | 76964 |
Gene Name: | RIKEN cDNA 2610028H24 gene |
Gene Aliases: |
ORF67 |
Location: |
Chr.10:76449023-76461218 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610028H24Rik | NM_029816.2 | NP_084092.2 | ||
| AK011591.1 | BAB27720.1 | |||
| AK161552.1 | BAE36458.1 | |||
| BC051454.1 | AAH51454.1 | |||
| BQ714071.1 |

Genomic Map