Assay Details
Target Gene Details
Entrez Gene ID: | 11567 |
Gene Name: | advillin |
Gene Aliases: |
Advil, DOC6 |
Location: |
Chr.10:127000075-127020994 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avil | NM_009635.3 | NP_033765.2 | ||
| AF041448.1 | AAC25050.1 | |||
| AF059486.1 | AAC31808.1 | |||
| AK154851.1 | BAE32877.1 | |||
| AK155900.1 | BAE33492.1 | |||
| AK171319.1 | BAE42389.1 | |||
| BC119222.1 | ||||
| BC120545.1 |
More Information
Set Membership: |
|

Genomic Map