Assay Details
Target Gene Details
Entrez Gene ID: | 27403 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 7 |
Gene Aliases: |
ABCX, Abc51 |
Location: |
Chr.10:79996453-80015572 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca7 | NM_013850.1 | NP_038878.1 | ||
| AF287141.1 | AAK56862.1 | |||
| BC145120.1 | ||||
| BC150708.1 |
More Information
Set Membership: |
|

Genomic Map