Assay Details
Target Gene Details
Entrez Gene ID: | 215748 |
Gene Name: | Cnksr family member 3 |
Gene Aliases: |
6820402C05, BC024086, Magi1 |
Location: |
Chr.10:7119061-7212237 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnksr3 | NM_172546.2 | 12 | 1876 | NP_766134.1 |
| AK033015.1 | 12 | 1877 | BAC28127.1 | |
| AK149951.1 | 11 | 1175 | BAE29189.1 | |
| AY151137.1 | 12 | 1838 | AAN72835.1 | |
| BC031194.1 | 5 | 983 | AAH31194.1 |

Genomic Map