Assay Details
Target Gene Details
Entrez Gene ID: | 68281 |
Gene Name: | RIKEN cDNA 4930430F08 gene |
Gene Aliases: |
4930571E09Rik, A730088G13Rik, AW551239, C430008C19Rik |
Location: |
Chr.10:100572274-100589390 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930430F08Rik | NM_175128.2 | NP_780337.2 | ||
| AK015251.1 | ||||
| AK032226.1 | BAC27770.1 | |||
| AK036647.1 | BAE43296.1 | |||
| AK043358.1 | 4 | 3239 | ||
| AK077761.1 | ||||
| AK078231.1 | BAC37184.1 | |||
| AK082894.1 | BAC38673.1 | |||
| AK160693.1 | ||||
| BC082587.1 | AAH82587.1 |

Genomic Map