Assay Details
Target Gene Details
Entrez Gene ID: | 83397 |
Gene Name: | A kinase (PRKA) anchor protein (gravin) 12 |
Gene Aliases: |
AI317366, SSeCKS, Srcs5, Tsga12 |
Location: |
Chr.10:4266329-4359471 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akap12 | NM_031185.3 | 2 | 400 | NP_112462.1 |
| AB020886.1 | 2 | 349 | BAA76894.1 | |
| AB025278.1 | 2 | 363 | BAC66465.1 | |
| AB051563.1 | 2 | 351 | BAB72161.1 | |
| AF326228.1 | 2 | 349 | AAK16150.1 | |
| AK053844.1 | 2 | 366 | BAC35553.1 | |
| BC071185.1 | 2 | 349 | AAH71185.1 | |
| BC138364.1 | 2 | 345 | ||
| BC138365.1 | 2 | 344 | ||
| CD540754.1 | 2 | 400 |
More Information
Set Membership: |
|

Genomic Map