Assay Details
Target Gene Details
Entrez Gene ID: | 53599 |
Gene Name: | CD164 antigen |
Gene Aliases: |
A115, A24, AA589639, AW540279, MGC-24, MSSP |
Location: |
Chr.10:41519500-41531042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd164 | NM_016898.2 | NP_058594.1 | ||
| AB014464.1 | BAA34547.1 | |||
| AB028895.1 | BAA78909.1 | |||
| AF299345.1 | AAG53910.1 | |||
| AK006592.1 | BAB24664.1 | |||
| AK018009.1 | ||||
| AK145131.1 | BAE26251.1 | |||
| AK169275.1 | BAE41034.1 | |||
| AK194033.1 | ||||
| BC005414.1 | AAH05414.1 |

Genomic Map