Assay Details
Target Gene Details
Entrez Gene ID: | 380660 |
Gene Name: | acyl-CoA synthetase short-chain family member 3 |
Gene Aliases: |
8430416H19Rik, Gm874 |
Location: |
Chr.10:106936164-107123664 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acss3 | NM_001142804.1 | 3 | 575 | NP_001136276.1 |
| NM_198636.3 | 3 | 575 | NP_941038.2 | |
| AK033376.1 | 3 | 580 | BAC28254.1 | |
| BC113198.1 | 3 | 539 | ||
| BY720548.1 | 3 | 575 |
More Information
Set Membership: |
|

Genomic Map